Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000745.3 | NP_000736.2 | 5 | 1395 | ||
GenBank mRNA | AF385586.1 | - | 5 | 1341 | ||
BC033639.1 | - | 5 | 1357 | 5501542 | ||
M83712.1 | - | 5 | 1343 | |||
Y08419.1 | - | 5 | 1195 |