Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198862.1 | NP_001185791.1 | 4 | 606 | ||
NM_018252.2 | NP_060722.2 | 3 | 447 | |||
XM_005273177.3 | XP_005273234.1 | 3 | 907 | |||
XM_011509717.2 | XP_011508019.1 | 3 | 706 | |||
XM_011509718.2 | XP_011508020.1 | 3 | 736 | |||
XM_017001636.1 | XP_016857125.1 | 2 | 290 | |||
XM_017001637.1 | XP_016857126.1 | 3 | 1030 | |||
GenBank mRNA | AK001736.1 | - | 3 | 417 | ||
AK024066.1 | - | 3 | 425 | |||
AK295224.1 | - | 3 | 437 | |||
AK297200.1 | - | 4 | 606 | |||
BC006320.2 | - | 3 | 406 | 4137890 | ||
CR457268.1 | - | 3 | 279 | |||
DC411913.1 | - | 3 | 447 |