Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000747.2 | NP_000738.2 | 3, 4 | 276 | ||
GenBank mRNA | AK298938.1 | - | 2, 3 | 130 | ||
AK308106.1 | - | 3, 4 | 265 | |||
BC011371.1 | - | 3, 4 | 264 | 3874415 | ||
BC023553.2 | - | 3, 4 | 276 | 3531876 | ||
X14830.1 | - | 3, 4 | 251 |