Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242875.2 | NP_001229804.1 | 14 | 1595 | ||
NM_001242876.2 | NP_001229805.1 | 12 | 1385 | |||
NM_001242877.2 | NP_001229806.1 | 12 | 1322 | |||
NM_001242878.2 | NP_001229807.1 | 11 | 1190 | |||
NM_001242879.2 | NP_001229808.1 | 10 | 1190 | |||
NM_001324465.1 | NP_001311394.1 | 12 | 1268 | |||
NM_001324466.1 | NP_001311395.1 | 13 | 1517 | |||
NM_001324467.1 | NP_001311396.1 | 12 | 1250 | |||
NM_001324468.1 | NP_001311397.1 | 12 | 1321 | |||
NM_018255.3 | NP_060725.1 | 13 | 1400 | |||
NR_040110.2 | 13 | 1400 | ||||
NR_136897.1 | 12 | 1333 | ||||
NR_136898.1 | 13 | 1400 | ||||
NR_137173.1 | 13 | 1400 | ||||
GenBank mRNA | AF332505.1 | - | 11 | 1194 | ||
AF403223.1 | - | 13 | 1377 | |||
AK001741.1 | - | 13 | 1350 | |||
AK022709.1 | - | 12 | 1326 | |||
AK222791.1 | - | 13 | 1353 | |||
AK293051.1 | - | 10 | 1156 | |||
AK300201.1 | - | 12 | 1339 | |||
AK302062.1 | - | 14 | 1561 | |||
BC032553.1 | - | 10 | 1155 | 5555577 | ||
CR457269.1 | - | 13 | 1340 |