Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242875.2 | NP_001229804.1 | 23 | 2664 | ||
NM_001242876.2 | NP_001229805.1 | 21 | 2454 | |||
NM_001242877.2 | NP_001229806.1 | 21 | 2391 | |||
NM_001242878.2 | NP_001229807.1 | 20 | 2259 | |||
NM_001242879.2 | NP_001229808.1 | 19 | 2259 | |||
NM_001324465.1 | NP_001311394.1 | 21 | 2337 | |||
NM_001324466.1 | NP_001311395.1 | 22 | 2586 | |||
NM_001324467.1 | NP_001311396.1 | 21 | 2319 | |||
NM_001324468.1 | NP_001311397.1 | 21 | 2390 | |||
NM_018255.3 | NP_060725.1 | 22 | 2469 | |||
NR_040110.2 | 22 | 2417 | ||||
NR_136897.1 | 21 | 2350 | ||||
NR_136898.1 | 22 | 2414 | ||||
NR_137173.1 | 21 | 2335 | ||||
GenBank mRNA | AF332505.1 | - | 20 | 2263 | ||
AF403223.1 | - | 22 | 2394 | |||
AK001741.1 | - | 22 | 2419 | |||
AK022709.1 | - | 21 | 2395 | |||
AK222791.1 | - | 22 | 2422 | |||
AK293051.1 | - | 19 | 2225 | |||
AK300201.1 | - | 21 | 2408 | |||
AK302062.1 | - | 23 | 2630 | |||
BC009211.2 | - | 7 | 759 | |||
BC032553.1 | - | 19 | 2224 | 5555577 | ||
CR457269.1 | - | 22 | 2409 |