Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCMTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104925.1 | NP_001098395.1 | 4 | 685 | ||
NM_018257.2 | NP_060727.2 | 4 | 685 | |||
GenBank mRNA | AK001745.1 | - | 4 | 612 | ||
BC032332.1 | - | 4 | 636 | 5169056 | ||
BC033665.2 | - | 3 | 510 | 4999114 | ||
BC040945.1 | - | 4 | 639 | 5743937 | ||
BC045775.1 | - | 4 | 476 | 4823937 |