Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000747.2 | NP_000738.2 | 8 | 953 | ||
GenBank mRNA | AK294880.1 | - | 5 | 1850 | ||
AK298938.1 | - | 7 | 807 | |||
AK308106.1 | - | 8 | 942 | |||
BC011371.1 | - | 8 | 941 | 3874415 | ||
BC023553.2 | - | 8 | 953 | 3531876 | ||
X14830.1 | - | 8 | 928 |