Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303538.1 | NP_001290467.1 | 4 | 789 | ||
NM_001303539.1 | NP_001290468.1 | 4 | 699 | |||
NM_001303540.1 | NP_001290469.1 | 3 | 594 | |||
NM_018273.3 | NP_060743.2 | 5 | 894 | |||
NR_130317.1 | 3 | 548 | ||||
GenBank mRNA | AK129801.1 | - | 5 | 597 | ||
AK291521.1 | - | 3 | 295 | |||
AK294408.1 | - | 3 | 295 | |||
AK297591.1 | - | 3 | 249 | |||
AK297610.1 | - | 4 | 490 | |||
AK298317.1 | - | 4 | 400 | |||
AK303635.1 | - | 4 | 402 | |||
AY358161.1 | - | 3 | 286 | |||
BC016919.1 | - | 5 | 586 | 4400601 | ||
BC040021.1 | - | 5 | 592 |