Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303538.1 | NP_001290467.1 | 5 | 1110 | ||
NM_001303539.1 | NP_001290468.1 | 5 | 1020 | |||
NM_001303540.1 | NP_001290469.1 | 4 | 915 | |||
NM_018273.3 | NP_060743.2 | 6 | 1215 | |||
NR_130317.1 | 4 | 869 | ||||
GenBank mRNA | AK129801.1 | - | 6 | 918 | ||
AK291521.1 | - | 4 | 616 | |||
AK297591.1 | - | 4 | 570 | |||
AK297610.1 | - | 5 | 811 | |||
AK298317.1 | - | 5 | 721 | |||
AY358161.1 | - | 4 | 607 | |||
BC016919.1 | - | 6 | 907 | 4400601 | ||
BC040021.1 | - | 6 | 913 |