Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303470.1 | NP_001290399.1 | 8 | 899 | ||
NM_018275.4 | NP_060745.3 | 8 | 1399 | |||
GenBank mRNA | AK001787.1 | - | 4 | 379 | ||
AK054872.1 | - | 8 | 895 | |||
AK302342.1 | - | 6 | 747 | |||
BC002749.2 | - | 8 | 799 | |||
BC015722.2 | - | 8 | 1357 | 3920412 |