Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGGY SIRNA Assays ›
Gene Symbol:
FGGY
Gene Name:
FGGY carbohydrate kinase domain containing
Gene Aliases:
-
Chromosome Location:
Chr.1: 59296405 - 59762730 on Build GRCh38
UniGene ID:
Hs.444301
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30617
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGGY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113411.1
NP_001106882.1
14
1625
NM_001244714.1
NP_001231643.1
11
1190
NM_001278224.1
NP_001265153.1
7
709
NM_018291.3
NP_060761.3
13
1553
NR_103473.1
14
1511
XM_011541730.1
XP_011540032.1
14
1549
XM_011541731.1
XP_011540033.1
14
1483
XM_011541732.1
XP_011540034.1
14
1526
XM_011541733.1
XP_011540035.1
14
1444
XM_011541735.1
XP_011540037.1
13
1372
XM_011541736.2
XP_011540038.1
12
1390
XM_017001643.1
XP_016857132.1
14
1655
XM_017001644.1
XP_016857133.1
14
1448
XM_017001645.1
XP_016857134.1
13
1477
XM_017001646.1
XP_016857135.1
13
1454
XM_017001647.1
XP_016857136.1
13
1582
XM_017001648.1
XP_016857137.1
15
1468
XM_017001649.1
XP_016857138.1
14
1594
XM_017001650.1
XP_016857139.1
15
1745
XM_017001651.1
XP_016857140.1
15
1601
XM_017001652.1
XP_016857141.1
13
1522
XM_017001653.1
XP_016857142.1
13
1454
XM_017001654.1
XP_016857143.1
12
1345
XM_017001655.1
XP_016857144.1
12
1254
XM_017001656.1
XP_016857145.1
15
1661
XM_017001657.1
XP_016857146.1
14
1449
XM_017001658.1
XP_016857147.1
15
1592
XM_017001659.1
XP_016857148.1
13
1482
XM_017001660.1
XP_016857149.1
13
1385
XM_017001661.1
XP_016857150.1
13
1542
XM_017001662.1
XP_016857151.1
13
1227
XM_017001663.1
XP_016857152.1
13
1402
XM_017001664.1
XP_016857153.1
12
1279
XM_017001665.1
XP_016857154.1
12
1186
XM_017001666.1
XP_016857155.1
11
1268
XM_017001667.1
XP_016857156.1
11
1322
XM_017001668.1
XP_016857157.1
12
1150
XM_017001669.1
XP_016857158.1
12
1245
XM_017001670.1
XP_016857159.1
12
2018
XM_017001671.1
XP_016857160.1
12
1146
XM_017001672.1
XP_016857161.1
12
1410
XM_017001673.1
XP_016857162.1
12
1467
XM_017001674.1
XP_016857163.1
12
1326
XM_017001675.1
XP_016857164.1
13
1482
XM_017001676.1
XP_016857165.1
12
1470
XM_017001677.1
XP_016857166.1
10
3720
XM_017001678.1
XP_016857167.1
10
4995
GenBank mRNA
AK001848.1
-
7
583
AK054842.1
-
11
1181
AK090568.1
-
14
1511
AK315649.1
-
13
1438
BC000610.2
-
7
684
BC014947.1
-
13
1438
4871664
BX647626.1
-
14
1597
Back To Top
More Information
Panther Classification:
Molecular Function -
carbohydrate kinase
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
phosphorylation
neuron cellular homeostasis
molecular_function
kinase activity
phosphotransferase activity, alcohol group as acceptor
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline