Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000749.3 | NP_000740.1 | 5 | 738 | ||
XM_011544390.1 | XP_011542692.1 | 3 | 561 | |||
GenBank mRNA | Y08417.1 | - | 5 | 738 |