Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018293.2 | NP_060763.2 | 1 | 1370 | ||
XM_011534340.2 | XP_011532642.2 | 7 | 2774 | |||
XM_017006789.1 | XP_016862278.1 | 8 | 2903 | |||
XM_017006790.1 | XP_016862279.1 | 8 | 2915 | |||
XM_017006791.1 | XP_016862280.1 | 6 | 2554 | |||
XM_017006792.1 | XP_016862281.1 | 6 | 2555 | |||
XM_017006793.1 | XP_016862282.1 | 7 | 2774 | |||
XM_017006794.1 | XP_016862283.1 | 6 | 2514 | |||
XM_017006795.1 | XP_016862284.1 | 7 | 2708 | |||
GenBank mRNA | AF543494.1 | - | 1 | 1371 | ||
AK001859.1 | - | 1 | 1157 | |||
AK024795.1 | - | 1 | 33 |