Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWF19L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303404.1 | NP_001290333.1 | 5 | 469 | ||
NM_001303405.1 | NP_001290334.1 | 5 | 514 | |||
NM_001303406.1 | NP_001290335.1 | 2 | 203 | |||
NM_018294.5 | NP_060764.3 | 5 | 469 | |||
GenBank mRNA | AK001860.1 | - | 5 | 415 | ||
AK225590.1 | - | 5 | 427 | |||
AK295303.1 | - | 2 | 173 | |||
AL832515.1 | - | 5 | 426 | |||
BC008746.2 | - | 5 | 410 | 3345869 | ||
DC299678.1 | - | 5 | 469 | |||
KU178692.1 | - | 5 | 381 | |||
KU178693.1 | - | 5 | 365 |