Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2W | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001481.2 | NP_001001481.2 | 5 | 391 | ||
NM_001271015.1 | NP_001257944.1 | 4 | 358 | |||
NM_018299.4 | NP_060769.4 | 4 | 358 | |||
NR_073119.1 | 3 | 255 | ||||
NR_073120.1 | 3 | 255 | ||||
NR_073121.1 | 3 | 255 | ||||
GenBank mRNA | AK001873.1 | - | 4 | 273 | ||
AK021735.1 | - | 4 | 273 | |||
AK024050.1 | - | 5 | 356 | |||
AK225554.1 | - | 4 | 289 | |||
AK295792.1 | - | 4 | 358 | |||
AY948289.1 | - | 5 | 327 | |||
BC010900.1 | - | 4 | 277 | 4081461 | ||
BC016326.1 | - | 4 | 312 | 4093131 | ||
BU155172.1 | - | 3 | 201 | |||
BX538145.1 | - | 3 | 178 | |||
BX648427.1 | - | 3 | 184 | |||
CR457275.1 | - | 3 | 270 | |||
DA352184.1 | - | 3 | 208 |