Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2W | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001481.2 | NP_001001481.2 | 3, 4 | 236 | ||
NM_001271015.1 | NP_001257944.1 | 2, 3 | 203 | |||
NM_018299.4 | NP_060769.4 | 2, 3 | 203 | |||
GenBank mRNA | AK001873.1 | - | 2, 3 | 118 | ||
AK021735.1 | - | 2, 3 | 118 | |||
AK024050.1 | - | 3, 4 | 201 | |||
AK225554.1 | - | 2, 3 | 134 | |||
AK295792.1 | - | 2, 3 | 203 | |||
AY948289.1 | - | 3, 4 | 172 | |||
BC010900.1 | - | 2, 3 | 122 | 4081461 | ||
BC016326.1 | - | 2, 3 | 157 | 4093131 | ||
CR457275.1 | - | 1, 2 | 115 |