Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000750.3 | NP_000741.1 | 2 | 225 | ||
NM_001256567.1 | NP_001243496.1 | 2 | 225 | |||
XM_011521186.2 | XP_011519488.1 | 4 | 255 | |||
XM_011521187.2 | XP_011519489.1 | 3 | 162 | |||
XM_011521192.2 | XP_011519494.1 | 2 | 218 | |||
XM_017021885.1 | XP_016877374.1 | 2 | 320 | |||
XM_017021886.1 | XP_016877375.1 | 2 | 2127 | |||
XM_017021887.1 | XP_016877376.1 | 2 | 129 | |||
XM_017021888.1 | XP_016877377.1 | 2 | 129 | |||
XM_017021889.1 | XP_016877378.1 | 2 | 129 | |||
GenBank mRNA | AK310443.1 | - | 8 | 975 | ||
BC096080.1 | - | 2 | 228 | 40000733 | ||
BC096081.1 | - | 2 | 228 | |||
BC096082.1 | - | 2 | 228 | 40000737 | ||
BC096083.1 | - | 2 | 228 | |||
U48861.1 | - | 2 | 233 | |||
U62439.1 | - | 2 | 199 | |||
Y08416.1 | - | 2 | 204 |