Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039702.2 | NP_001034791.1 | 5 | 617 | ||
NM_018324.2 | NP_060794.1 | 6 | 776 | |||
XM_017016376.1 | XP_016871865.1 | 6 | 740 | |||
GenBank mRNA | AK001844.1 | - | 5 | 567 | ||
AK001968.1 | - | 6 | 709 | |||
BC050372.1 | - | 5 | 584 | 5742377 | ||
BG722790.1 | - | 5 | 617 | |||
BI830605.1 | - | 5 | 620 |