Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIMAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018326.2 | NP_060796.1 | 3 | 582 | ||
XM_005250017.1 | XP_005250074.1 | 3 | 598 | |||
GenBank mRNA | AF117333.1 | - | 3 | 550 | ||
AK001972.1 | - | 3 | 544 | |||
AK301441.1 | - | 3 | 601 | |||
AK301606.1 | - | 4 | 350 | |||
BC020657.1 | - | 3 | 559 | 4701647 |