Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX19A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320522.1 | NP_001307451.1 | 2, 3 | 277 | ||
NM_001320525.1 | NP_001307454.1 | 2, 3 | 277 | |||
NM_001320526.1 | NP_001307455.1 | 2, 3 | 277 | |||
NM_001320527.1 | NP_001307456.1 | 2, 3 | 277 | |||
NM_018332.4 | NP_060802.1 | 2, 3 | 277 | |||
NR_135296.1 | 2, 3 | 277 | ||||
GenBank mRNA | AK001988.1 | - | 2, 3 | 235 | ||
AK223262.1 | - | 2, 3 | 201 | |||
AK299539.1 | - | 2, 3 | 162 | |||
AK310604.1 | - | 2, 3 | 201 | |||
AK310804.1 | - | 2, 3 | 203 | |||
AL832970.1 | - | 2, 3 | 188 | |||
BC005162.2 | - | 2, 3 | 185 | 3530064 | ||
BC006544.2 | - | 2, 3 | 185 | 2964377 | ||
BC137496.1 | - | 2, 3 | 163 | 9021496 | ||
BC137497.1 | - | 2, 3 | 163 | 9021497 | ||
CR749227.1 | - | 2, 3 | 224 | |||
DB074731.1 | - | 2, 3 | 277 | |||
DC375258.1 | - | 2, 3 | 245 |