Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000080.3 | NP_000071.1 | 2, 3 | 198 | ||
XM_017024115.1 | XP_016879604.1 | 2, 3 | 217 | |||
GenBank mRNA | X66403.1 | - | 2, 3 | 198 |