Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018346.2 | NP_060816.1 | 3 | 483 | ||
GenBank mRNA | AK002026.1 | - | 3 | 454 | ||
AK022105.1 | - | 3 | 423 | |||
AK222472.1 | - | 3 | 434 | |||
AK293814.1 | - | 3 | 423 | |||
BC005854.2 | - | 2 | 235 | |||
BC050538.1 | - | 3 | 414 | 5214885 | ||
DC370821.1 | - | 3 | 488 |