Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018358.2 | NP_060828.2 | 13, 14 | 1455 | ||
XM_011512959.2 | XP_011511261.1 | 13, 14 | 1459 | |||
GenBank mRNA | AK002060.1 | - | 13, 14 | 1432 | ||
AK024758.1 | - | 7, 8 | 654 | |||
AK027288.1 | - | 7, 8 | 723 | |||
AK092415.1 | - | 13, 14 | 1338 | |||
AK097073.1 | - | 12, 13 | 1722 | |||
AK126150.1 | - | 1 | 33 | |||
AK290106.1 | - | 13, 14 | 1327 | |||
AK299433.1 | - | 13, 14 | 1219 | |||
BC009253.2 | - | 13, 14 | 1334 | 3028775 | ||
BC035652.1 | - | 13, 14 | 1329 | |||
BC051754.2 | - | 13, 14 | 1329 | 6064628 | ||
BC051884.1 | - | 13, 14 | 1329 | 5105761 | ||
KU178694.1 | - | 13, 14 | 1322 | |||
KU178695.1 | - | 13, 14 | 1279 |