Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018370.2 | NP_060840.2 | 1, 2 | 600 | ||
XM_005269004.2 | XP_005269061.1 | 1, 2 | 485 | |||
XM_005269005.2 | XP_005269062.1 | 1, 2 | 600 | |||
GenBank mRNA | AK094923.1 | - | 1, 2 | 230 | ||
AK297785.1 | - | 1, 2 | 244 | |||
BC018435.1 | - | 1, 2 | 576 | 4474297 |