Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252148.1 | NP_001239077.1 | 2, 3 | 901 | ||
NM_001252150.1 | NP_001239079.1 | 2, 3 | 901 | |||
NM_001252151.1 | NP_001239080.1 | 2, 3 | 328 | |||
NM_018375.4 | NP_060845.2 | 2, 3 | 901 | |||
GenBank mRNA | AK002136.1 | - | 2, 3 | 877 | ||
AK222547.1 | - | 2, 3 | 879 | |||
AK293222.1 | - | 2, 3 | 321 | |||
AY358687.1 | - | 2, 3 | 347 | |||
AY780789.1 | - | 2, 3 | 199 | |||
BC047682.1 | - | 2, 3 | 831 | 6066186 | ||
BC064383.1 | - | 2, 3 | 616 | 5737339 | ||
BP269907.1 | - | 1, 2 | 103 | |||
BX643103.1 | - | 2, 3 | 471 | |||
FJ423549.1 | - | 2, 3 | 199 |