Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPSNAP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018376.3 | NP_060846.2 | 4 | 1041 | ||
NR_130759.1 | 3 | 830 | ||||
XM_017014889.1 | XP_016870378.1 | 4 | 993 | |||
GenBank mRNA | AK002137.1 | - | 4 | 646 | ||
AK310397.1 | - | 3 | 386 | |||
BC005202.1 | - | 4 | 574 | 3933738 | ||
BC017914.1 | - | 4 | 612 | 4292375 | ||
BX647652.1 | - | 4 | 613 |