Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018397.4 | NP_060867.2 | 9 | 2128 | ||
XM_006713250.3 | XP_006713313.1 | 9 | 1910 | |||
XM_006713251.3 | XP_006713314.1 | 9 | 2087 | |||
XM_006713252.3 | XP_006713315.1 | 8 | 1767 | |||
XM_011533938.2 | XP_011532240.1 | 9 | 1944 | |||
XM_011533939.2 | XP_011532241.1 | 9 | 2783 | |||
XM_017006797.1 | XP_016862286.1 | 9 | 1941 | |||
XM_017006798.1 | XP_016862287.1 | 9 | 1933 | |||
XM_017006799.1 | XP_016862288.1 | 8 | 1764 | |||
GenBank mRNA | AJ272267.1 | - | 7 | 1352 | ||
AK055402.1 | - | 1 | 1516 | |||
AK297575.1 | - | 5 | 811 | |||
BC034502.1 | - | 9 | 1886 | 4824572 |