Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAPTM4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018407.4 | NP_060877.3 | 5 | 888 | ||
GenBank mRNA | AF317417.1 | - | 5 | 459 | ||
AF527412.1 | - | 5 | 817 | |||
AK075326.1 | - | 5 | 650 | |||
AL136942.1 | - | 5 | 642 | |||
AY057051.2 | - | 5 | 888 | |||
AY261384.1 | - | 5 | 907 | |||
BC014129.2 | - | 5 | 644 | 4581284 | ||
BC031021.1 | - | 5 | 669 | 4690368 | ||
BC038117.1 | - | 5 | 1436 | 5264567 |