Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018420.2 | NP_060890.2 | 5 | 775 | ||
XM_005271004.3 | XP_005271061.1 | 5 | 736 | |||
GenBank mRNA | AK125072.1 | - | 3 | 343 | ||
AY145501.1 | - | 5 | 645 | |||
AY358258.1 | - | 5 | 659 | |||
BC026358.1 | - | 5 | 774 | 4814675 | ||
KU178698.1 | - | 5 | 609 | |||
KU178700.1 | - | 5 | 609 | |||
KU178701.1 | - | 4 | 396 | |||
KU178703.1 | - | 4 | 396 |