Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STYK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018423.2 | NP_060893.2 | 4 | 617 | ||
XM_005253417.2 | XP_005253474.1 | 3 | 517 | |||
XM_011520736.1 | XP_011519038.1 | 5 | 420 | |||
XM_011520737.2 | XP_011519039.1 | 4 | 457 | |||
XM_011520738.2 | XP_011519040.1 | 4 | 612 | |||
XM_017019579.1 | XP_016875068.1 | 4 | 425 | |||
GenBank mRNA | AF251059.1 | - | 4 | 617 | ||
AK313906.1 | - | 4 | 352 | |||
AL353940.1 | - | 4 | 365 | |||
AY563054.1 | - | 2 | 96 | |||
BC093822.1 | - | 2 | 118 | 7939667 | ||
BC093824.1 | - | 2 | 118 | 7939669 | ||
KP729000.1 | - | 2 | 96 |