Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI4K2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018425.3 | NP_060895.1 | 7 | 1221 | ||
GenBank mRNA | AJ303098.1 | - | 7 | 1221 | ||
AL353952.1 | - | 5 | 502 | |||
BC003167.1 | - | 7 | 1193 | 2905670 | ||
BT007330.1 | - | 7 | 1164 |