Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM63B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318792.1 | NP_001305721.1 | 20 | 2033 | ||
NM_018426.2 | NP_060896.1 | 20 | 1999 | |||
XM_005249211.3 | XP_005249268.1 | 22 | 2379 | |||
XM_005249213.4 | XP_005249270.1 | 21 | 2306 | |||
XM_005249214.4 | XP_005249271.1 | 19 | 1893 | |||
XM_005249215.4 | XP_005249272.1 | 20 | 2198 | |||
XM_005249217.1 | XP_005249274.1 | 20 | 2071 | |||
XM_006715135.1 | XP_006715198.1 | 20 | 2055 | |||
XM_017010999.1 | XP_016866488.1 | 22 | 2256 | |||
XM_017011000.1 | XP_016866489.1 | 21 | 2237 | |||
XM_017011001.1 | XP_016866490.1 | 19 | 1937 | |||
XM_017011002.1 | XP_016866491.1 | 19 | 2040 | |||
GenBank mRNA | AL353957.1 | - | 6 | 457 | ||
AL834238.1 | - | 13 | 1297 | |||
BC022095.1 | - | 17 | 1636 | |||
BC062989.1 | - | 8 | 825 | |||
BC136769.1 | - | 19 | 1875 | 9020759 | ||
CR627460.1 | - | 20 | 2036 |