Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM63B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318792.1 | NP_001305721.1 | 5 | 529 | ||
NM_018426.2 | NP_060896.1 | 5 | 495 | |||
XM_005249211.3 | XP_005249268.1 | 7 | 875 | |||
XM_005249213.4 | XP_005249270.1 | 6 | 802 | |||
XM_005249214.4 | XP_005249271.1 | 4 | 389 | |||
XM_005249215.4 | XP_005249272.1 | 5 | 694 | |||
XM_005249217.1 | XP_005249274.1 | 5 | 567 | |||
XM_006715135.1 | XP_006715198.1 | 5 | 551 | |||
XM_017010999.1 | XP_016866488.1 | 7 | 752 | |||
XM_017011000.1 | XP_016866489.1 | 6 | 733 | |||
XM_017011001.1 | XP_016866490.1 | 4 | 433 | |||
XM_017011002.1 | XP_016866491.1 | 4 | 536 | |||
GenBank mRNA | AK093629.1 | - | 5 | 551 | ||
AK309791.1 | - | 3 | 398 | |||
BC022095.1 | - | 2 | 132 | |||
BC136769.1 | - | 4 | 371 | 9020759 | ||
CR627460.1 | - | 5 | 532 | |||
DC408120.1 | - | 5 | 495 |