Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300980.1 | NP_001287909.1 | 5 | 633 | ||
NM_018502.4 | NP_060972.3 | 5 | 633 | |||
XM_005268477.1 | XP_005268534.1 | 5 | 633 | |||
XM_011537663.2 | XP_011535965.1 | 5 | 633 | |||
XM_011537665.2 | XP_011535967.1 | 4 | 697 | |||
XM_017009617.1 | XP_016865106.1 | 5 | 633 | |||
XM_017009618.1 | XP_016865107.1 | 5 | 633 | |||
XM_017009619.1 | XP_016865108.1 | 4 | 421 | |||
XM_017009620.1 | XP_016865109.1 | 4 | 424 | |||
GenBank mRNA | AK097088.1 | - | 5 | 597 | ||
AK301502.1 | - | 3 | 957 | |||
AK308722.1 | - | 3 | 953 | |||
BC001910.1 | - | 5 | 556 | 3538001 | ||
BQ066786.1 | - | 5 | 621 |