Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIRBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280.2 | NP_001271.1 | 2 | 240 | ||
NM_001300815.1 | NP_001287744.1 | 2 | 242 | |||
NM_001300829.1 | NP_001287758.1 | 2 | 242 | |||
NR_023312.2 | 2 | 301 | ||||
NR_023313.2 | 2 | 240 | ||||
XM_006722637.1 | XP_006722700.1 | 2 | 303 | |||
XM_011527668.1 | XP_011525970.1 | 2 | 176 | |||
XM_017026237.1 | XP_016881726.1 | 2 | 303 | |||
GenBank mRNA | AB451225.1 | - | 1 | 91 | ||
AB451348.1 | - | 1 | 91 | |||
AF021336.1 | - | 2 | 173 | |||
AK094781.1 | - | 2 | 173 | |||
AK095781.1 | - | 2 | 176 | |||
AK128423.1 | - | 2 | 175 | |||
AK304185.1 | - | 2 | 237 | |||
AK304470.1 | - | 2 | 176 | |||
AK311601.1 | - | 2 | 176 | |||
BC000403.2 | - | 2 | 152 | 2822968 | ||
BC000901.1 | - | 2 | 157 | 2900585 | ||
BM706055.1 | - | 2 | 154 | |||
BP195090.1 | - | 2 | 240 | |||
BT007210.1 | - | 1 | 91 | |||
D78134.1 | - | 2 | 171 | |||
DA433715.1 | - | 2 | 177 | |||
DA695021.1 | - | 2 | 304 |