Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083946.1 | NP_001077415.1 | 6 | 638 | ||
NM_144736.4 | NP_653337.1 | 8 | 932 | |||
XM_005264410.1 | XP_005264467.1 | 8 | 912 | |||
XM_005264411.1 | XP_005264468.1 | 7 | 697 | |||
XM_011532956.1 | XP_011531258.1 | 8 | 868 | |||
GenBank mRNA | AK299015.1 | - | 7 | 693 | ||
AK302150.1 | - | 7 | 802 | |||
AK303555.1 | - | 7 | 821 | |||
BC004548.2 | - | 8 | 897 | 3959139 | ||
BC012374.1 | - | 3 | 246 | |||
BX538031.1 | - | 6 | 591 |