Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CISH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013324.5 | NP_037456.5 | 4 | 651 | ||
NM_145071.2 | NP_659508.1 | 3 | 494 | |||
XM_011533329.2 | XP_011531631.1 | 2 | 3316 | |||
GenBank mRNA | AF035947.1 | - | 2 | 3462 | ||
AF132297.2 | - | 4 | 592 | |||
AK313850.1 | - | 3 | 437 | |||
BC031590.1 | - | 3 | 426 | 5182644 | ||
BC064354.1 | - | 3 | 331 | 4804249 | ||
D83532.1 | - | 3 | 424 | |||
DA957793.1 | - | 3 | 494 |