Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018648.3 | NP_061118.1 | 1 | 130 | ||
GenBank mRNA | AB043104.1 | - | 1 | 140 | ||
BC008886.2 | - | 1 | 75 | 4308721 | ||
BC063023.1 | - | 1 | 101 | 6018963 | ||
BM554889.1 | - | 1 | 71 | |||
BQ218536.1 | - | 1 | 146 |