Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018656.2 | NP_061126.2 | 3 | 759 | ||
XM_005269006.3 | XP_005269063.1 | 3 | 767 | |||
XM_017019584.1 | XP_016875073.1 | 2 | 656 | |||
GenBank mRNA | AF148713.1 | - | 3 | 731 | ||
AK289645.1 | - | 3 | 767 | |||
AY358943.1 | - | 3 | 759 | |||
BC008412.1 | - | 3 | 700 | 4280446 | ||
BC021103.1 | - | 3 | 710 | 4810919 | ||
BC030504.2 | - | 3 | 727 | 3900045 | ||
BC049192.1 | - | 3 | 710 | 3922727 |