Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018665.2 | NP_061135.2 | 1 | 446 | ||
XM_011535926.1 | XP_011534228.1 | 1 | 446 | |||
XM_011535928.2 | XP_011534230.1 | 1 | 148 | |||
XM_017011003.1 | XP_016866492.1 | 1 | 114 | |||
GenBank mRNA | AJ278110.1 | - | 1 | 309 | ||
AK303324.1 | - | 1 | 155 | |||
AK313686.1 | - | 1 | 146 | |||
AL136751.1 | - | 1 | 134 | |||
BC066938.1 | - | 1 | 127 | 4829072 | ||
BX646425.1 | - | 1 | 193 |