Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAGE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018666.2 | NP_061136.2 | 7 | 858 | ||
XM_017029621.1 | XP_016885110.1 | 7 | 866 | |||
XM_017029622.1 | XP_016885111.1 | 7 | 777 | |||
XM_017029623.1 | XP_016885112.1 | 7 | 773 | |||
XM_017029624.1 | XP_016885113.1 | 6 | 680 | |||
XM_017029625.1 | XP_016885114.1 | 7 | 773 | |||
GenBank mRNA | AJ278111.1 | - | 7 | 858 | ||
AK310412.1 | - | 6 | 814 |