Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182847.2 | NP_878267.2 | 5 | 1701 | ||
XM_005246671.2 | XP_005246728.2 | 6 | 1615 | |||
XM_017004439.1 | XP_016859928.1 | 6 | 774 | |||
GenBank mRNA | BC010439.1 | - | 5 | 1541 | 4215164 | |
EL736952.1 | - | 5 | 546 |