Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018696.2 | NP_061166.1 | 4 | 1037 | ||
GenBank mRNA | AB029151.1 | - | 4 | 977 | ||
AF308695.1 | - | 4 | 1021 | |||
BC014624.2 | - | 4 | 998 | 2960155 |