Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018725.3 | NP_061195.2 | 11 | 1046 | ||
XM_005265310.4 | XP_005265367.1 | 12 | 1133 | |||
XM_005265311.4 | XP_005265368.1 | 11 | 1110 | |||
XM_005265312.4 | XP_005265369.1 | 10 | 1054 | |||
XM_017006804.1 | XP_016862293.1 | 11 | 1002 | |||
XM_017006805.1 | XP_016862294.1 | 10 | 954 | |||
XM_017006806.1 | XP_016862295.1 | 10 | 915 | |||
GenBank mRNA | AF208110.1 | - | 11 | 1012 | ||
AF208111.1 | - | 10 | 2041 | |||
AF212365.1 | - | 11 | 1016 | |||
AF250309.1 | - | 11 | 1007 | |||
AK307749.1 | - | 10 | 1004 | |||
AY359122.1 | - | 11 | 1013 | |||
BC000980.1 | - | 11 | 1016 | 2985728 |