Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080493.3 | NP_001073962.1 | 4 | 465 | ||
NM_001297610.1 | NP_001284539.1 | 3 | 404 | |||
NM_017507.1 | NP_059977.1 | 3 | 338 | |||
XM_017026941.1 | XP_016882430.1 | 4 | 627 | |||
XM_017026942.1 | XP_016882431.1 | 2 | 265 | |||
GenBank mRNA | AK300579.1 | - | 4 | 432 | ||
AK303213.1 | - | 3 | 369 | |||
AK308189.1 | - | 3 | 274 | |||
BC052597.1 | - | 4 | 377 | 4304302 | ||
DA519419.1 | - | 3 | 404 | |||
DA880101.1 | - | 3 | 274 | |||
X51760.1 | - | 4 | 379 |