Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKMT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015001.2 | NP_001015001.1 | 4 | 917 | ||
NM_001321926.1 | NP_001308855.1 | 3 | 785 | |||
NM_001321927.1 | NP_001308856.1 | 4 | 1010 | |||
NM_001321928.1 | NP_001308857.1 | 4 | 771 | |||
NM_001321929.1 | NP_001308858.1 | 3 | 813 | |||
NR_135856.1 | 3 | 1062 | ||||
XM_005254498.3 | XP_005254555.1 | 3 | 536 | |||
XM_017022369.1 | XP_016877858.1 | 4 | 842 | |||
XM_017022370.1 | XP_016877859.1 | 4 | 767 | |||
XM_017022371.1 | XP_016877860.1 | 4 | 1152 | |||
GenBank mRNA | AK223365.1 | - | 3 | 597 | ||
AK295776.1 | - | 3 | 661 | |||
AK316052.1 | - | 4 | 461 | |||
AK316124.1 | - | 4 | 868 | |||
AK316319.1 | - | 4 | 771 | |||
BC001926.1 | - | 3 | 602 | 2820642 | ||
BC006467.1 | - | 3 | 528 | 3535486 | ||
BT006628.1 | - | 3 | 425 |
Gene Symbol: CKMT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020990.4 | NP_066270.1 | 4 | 1014 | ||
NR_135748.1 | 3 | 1063 | ||||
NR_135749.1 | 3 | 1087 | ||||
NR_135750.1 | 4 | 1014 | ||||
NR_135751.1 | 3 | 903 | ||||
XM_005254150.3 | XP_005254207.1 | 3 | 536 | |||
XM_011521194.1 | XP_011519496.1 | 4 | 1102 | |||
XM_011521195.2 | XP_011519497.1 | 4 | 841 | |||
XM_011521196.1 | XP_011519498.1 | 4 | 767 | |||
XM_011521197.2 | XP_011519499.1 | 3 | 601 | |||
XM_011521198.1 | XP_011519500.1 | 4 | 1286 | |||
XM_011521199.2 | XP_011519501.2 | 4 | 1102 | |||
XM_017021902.1 | XP_016877391.1 | 4 | 1286 | |||
GenBank mRNA | AK094322.1 | - | 4 | 592 | ||
AK293685.1 | - | 3 | 330 | |||
AK293790.1 | - | 3 | 870 | |||
AK293939.1 | - | 3 | 667 | |||
AK293961.1 | - | 2 | 452 | |||
AK294002.1 | - | 3 | 712 | |||
AK294735.1 | - | 4 | 485 | |||
AK294908.1 | - | 3 | 902 | |||
AK296271.1 | - | 2 | 147 | |||
AK296472.1 | - | 3 | 549 | |||
AK308770.1 | - | 2 | 419 | |||
BC108652.1 | - | 3 | 537 | 6190035 | ||
BC121001.1 | - | 3 | 601 | 40116549 | ||
BC121002.1 | - | 3 | 601 | 40116552 |