Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKMT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015001.2 | NP_001015001.1 | 5 | 975 | ||
NM_001321926.1 | NP_001308855.1 | 4 | 843 | |||
NM_001321927.1 | NP_001308856.1 | 5 | 1068 | |||
NM_001321928.1 | NP_001308857.1 | 5 | 829 | |||
XM_017022369.1 | XP_016877858.1 | 5 | 900 | |||
XM_017022370.1 | XP_016877859.1 | 5 | 825 | |||
GenBank mRNA | AK223365.1 | - | 4 | 655 | ||
AK295776.1 | - | 4 | 719 | |||
AK316124.1 | - | 5 | 926 | |||
AK316319.1 | - | 5 | 829 | |||
BC001926.1 | - | 4 | 660 | 2820642 | ||
BC006467.1 | - | 4 | 586 | 3535486 | ||
BT006628.1 | - | 4 | 483 |
Gene Symbol: CKMT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020990.4 | NP_066270.1 | 5 | 1072 | ||
NR_135748.1 | 4 | 1121 | ||||
NR_135750.1 | 5 | 1072 | ||||
NR_135751.1 | 4 | 961 | ||||
XM_011521194.1 | XP_011519496.1 | 5 | 1160 | |||
XM_011521195.2 | XP_011519497.1 | 5 | 899 | |||
XM_011521196.1 | XP_011519498.1 | 5 | 825 | |||
XM_011521197.2 | XP_011519499.1 | 4 | 659 | |||
XM_011521198.1 | XP_011519500.1 | 5 | 1344 | |||
GenBank mRNA | AK094322.1 | - | 5 | 650 | ||
AK293790.1 | - | 4 | 928 | |||
AK293939.1 | - | 4 | 725 | |||
AK293961.1 | - | 3 | 510 | |||
AK294735.1 | - | 5 | 543 | |||
AK294908.1 | - | 4 | 960 | |||
BC108652.1 | - | 4 | 595 | 6190035 | ||
BC121001.1 | - | 4 | 659 | 40116549 | ||
BC121002.1 | - | 4 | 659 | 40116552 |