Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017518.7 | NP_059988.3 | 3 | 354 | ||
GenBank mRNA | AF267739.1 | - | 3 | 285 | ||
AK296147.1 | - | 5 | 603 | |||
AK300650.1 | - | 3 | 306 | |||
AK302066.1 | - | 3 | 285 | |||
BC008141.1 | - | 3 | 354 | 4154976 |