Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201552.1 | NP_001188481.1 | 5 | 721 | ||
NM_001201553.1 | NP_001188482.1 | 4 | 706 | |||
NM_001201554.1 | NP_001188483.1 | 4 | 595 | |||
NM_001201556.1 | NP_001188485.1 | 5 | 721 | |||
NM_001318239.1 | NP_001305168.1 | 4 | 448 | |||
NM_017530.2 | NP_060000.1 | 3 | 580 | |||
XM_005256033.2 | XP_005256090.1 | 6 | 795 | |||
XM_006721233.1 | XP_006721296.1 | 5 | 654 | |||
XM_011523211.2 | XP_011521513.1 | 4 | 500 | |||
XM_017023409.1 | XP_016878898.1 | 5 | 639 | |||
XM_017023410.1 | XP_016878899.1 | 5 | 1386 | |||
XM_017023411.1 | XP_016878900.1 | 5 | 463 | |||
XM_017023412.1 | XP_016878901.1 | 6 | 795 | |||
XM_017023414.1 | XP_016878903.1 | 4 | 582 | |||
XM_017023416.1 | XP_016878905.1 | 5 | 732 | |||
XM_017023417.1 | XP_016878906.1 | 5 | 726 | |||
XM_017023418.1 | XP_016878907.1 | 5 | 588 | |||
XM_017023419.1 | XP_016878908.1 | 5 | 570 | |||
XM_017023420.1 | XP_016878909.1 | 6 | 815 | |||
XM_017023422.1 | XP_016878911.1 | 3 | 236 | |||
XM_017023423.1 | XP_016878912.1 | 5 | 729 | |||
XM_017023424.1 | XP_016878913.1 | 4 | 583 | |||
GenBank mRNA | AF070588.1 | - | 3 | 548 | ||
AK296603.1 | - | 4 | 706 | |||
AK313693.1 | - | 4 | 555 | |||
BC012116.1 | - | 4 | 552 | 4553333 |