Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF821 SIRNA Assays ›
Gene Symbol:
ZNF821
Gene Name:
zinc finger protein 821
Gene Aliases:
-
Chromosome Location:
Chr.16: 71859680 - 71886224 on Build GRCh38
UniGene ID:
Hs.744055
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30987
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF821
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001201552.1
NP_001188481.1
8
1205
NM_001201553.1
NP_001188482.1
7
1190
NM_001201554.1
NP_001188483.1
7
1079
NM_001201556.1
NP_001188485.1
7
1038
NM_001318238.1
NP_001305167.1
8
1159
NM_001318239.1
NP_001305168.1
7
932
NM_017530.2
NP_060000.1
6
1064
XM_005256033.2
XP_005256090.1
9
1279
XM_006721233.1
XP_006721296.1
8
1138
XM_011523211.2
XP_011521513.1
7
984
XM_011523212.2
XP_011521514.1
7
1037
XM_011523214.2
XP_011521516.1
7
1007
XM_011523215.2
XP_011521517.1
5
678
XM_011523220.2
XP_011521522.1
6
760
XM_017023409.1
XP_016878898.1
8
1123
XM_017023410.1
XP_016878899.1
8
1870
XM_017023411.1
XP_016878900.1
8
947
XM_017023412.1
XP_016878901.1
9
1279
XM_017023413.1
XP_016878902.1
7
1007
XM_017023414.1
XP_016878903.1
7
1066
XM_017023415.1
XP_016878904.1
7
1004
XM_017023416.1
XP_016878905.1
8
1216
XM_017023417.1
XP_016878906.1
8
1210
XM_017023418.1
XP_016878907.1
8
1072
XM_017023419.1
XP_016878908.1
8
1054
XM_017023420.1
XP_016878909.1
9
1299
XM_017023421.1
XP_016878910.1
7
1065
XM_017023422.1
XP_016878911.1
6
720
XM_017023423.1
XP_016878912.1
7
1046
XM_017023424.1
XP_016878913.1
6
900
XM_017023425.1
XP_016878914.1
5
754
GenBank mRNA
AF070588.1
-
6
1032
AK296603.1
-
7
1190
AK313693.1
-
7
1039
BC012116.1
-
7
1036
4553333
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline